Tisk…

5-Bromo-3-(1,2,3,6-tetrahydro-4-pyridinyl)-1H-indole 95%

Dodavatel: Apollo Scientific

OR32600-1G OR32600-5G
APOSOR32600-1GEA 9560 CZK
APOSOR32600-1G APOSOR32600-5G
5-Bromo-3-(1,2,3,6-tetrahydro-4-pyridinyl)-1H-indole 95%
5-Bromo-3-(1,2,3,6-tetrahydro-4-pyridinyl)-1H-indole
Vzorec: C₁₃H₁₃BrN₂
Storage Temperature: Ambient
MDL Number: MFCD07366543
CAS číslo: 127792-80-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR