Tisk…

1-Methyl-1H-pyrazolo[3,4-b]pyridin-3-ylamine 97%

Dodavatel: Apollo Scientific

OR32589-5G
APOSOR32589-5GEA 4040 CZK
APOSOR32589-5G
1-Methyl-1H-pyrazolo[3,4-b]pyridin-3-ylamine 97%
1-Methyl-1H-pyrazolo[3,4-b]pyridin-3-ylamine
Vzorec: C₇H₈N₄
MW: 148,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04117796
CAS číslo: 72583-83-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR