Tisk…

2-(1-Oxo-1,3-dihydro-2H-isoindol-2-yl)benzenecarboxylic acid 95%

Dodavatel: Apollo Scientific

Synonyms: 2-(1-Oxo-2,3-dihydro-1H-isoindol-2-yl)benzoic acid

OR32569-1G OR32569-5G
APOSOR32569-1GEA 9560 CZK
APOSOR32569-1G APOSOR32569-5G
2-(1-Oxo-1,3-dihydro-2H-isoindol-2-yl)benzenecarboxylic acid 95%
2-(1-Oxo-1,3-dihydro-2H-isoindol-2-yl)benzenecarboxylic acid
Vzorec: C₁₅H₁₁NO₃
MW: 253,26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02186096
CAS číslo: 4770-69-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR