Tisk…

Ethyl-2-[(4-chlorobenzoyl)amino]acetate 95%

Dodavatel: Apollo Scientific

OR32537-1G OR32537-5G
APOSOR32537-1GEA 5130 CZK
APOSOR32537-1G APOSOR32537-5G
Ethyl-2-[(4-chlorobenzoyl)amino]acetate 95%
Ethyl-2-[(4-chlorobenzoyl)amino]acetate
Vzorec: C₁₁H₁₂ClNO₃
Storage Temperature: Ambient
MDL Number: MFCD00095955
CAS číslo: 39735-52-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR