Tisk…

5-Chloro-4-phenyl-1,2,3-thiadiazole 95%

Dodavatel: Apollo Scientific

OR32534-1G OR32534-5G
APOSOR32534-1GEA 9560 CZK
APOSOR32534-1G APOSOR32534-5G
5-Chloro-4-phenyl-1,2,3-thiadiazole 95%
5-Chloro-4-phenyl-1,2,3-thiadiazole
Vzorec: C₈H₅ClN₂S
Storage Temperature: Ambient
MDL Number: MFCD00100225
CAS číslo: 53646-00-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR