Tisk…

2-Cyano-3',4'-dichloroacetanilide 95%

Dodavatel: Apollo Scientific

Synonyms: 2-Cyano-N-(3,4-dichlorophenyl)acetamide

OR32525-1G OR32525-25G OR32525-5G
APOSOR32525-1GEA 5130 CZK
APOSOR32525-1G APOSOR32525-25G APOSOR32525-5G
2-Cyano-3',4'-dichloroacetanilide 95%
2-Cyano-3',4'-dichloroacetanilide
Vzorec: C₉H₆Cl₂N₂O
Storage Temperature: Ambient
MDL Number: MFCD00036014
CAS číslo: 15386-80-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR