Tisk…

2-Methyl-3-(pyrrol-1-yl)aniline 95%

Dodavatel: Apollo Scientific

OR32493-1G OR32493-5G OR32493-25G
APOSOR32493-1GEA 3480 CZK
APOSOR32493-1G APOSOR32493-5G APOSOR32493-25G
2-Methyl-3-(pyrrol-1-yl)aniline 95%
2-Methyl-3-(pyrrol-1-yl)aniline
Vzorec: C₁₁H₁₂N₂
Storage Temperature: Ambient
CAS číslo: 137352-75-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR