Tisk…

2-Chloro-4-(1H-pyrazol-1-yl)benzenecarboxylic acid 95%

Dodavatel: Apollo Scientific

Synonyms: 2-Chloro-4-(1H-pyrazol-1-yl)benzoic acid

OR32485-1G OR32485-5G
APOSOR32485-1GEA 3940 CZK
APOSOR32485-1G APOSOR32485-5G
2-Chloro-4-(1H-pyrazol-1-yl)benzenecarboxylic acid 95%
2-Chloro-4-(1H-pyrazol-1-yl)benzenecarboxylic acid
Vzorec: C₁₀H₇ClN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD03425733
CAS číslo: 313674-12-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR