Tisk…
Methyl-2-chloro-4-(1-pyrrolidinyl)benzenecarboxylate 95%
Dodavatel: Apollo Scientific
Synonyms: Methyl 2-Chloro-4-(1-pyrrolidinyl)benzoate, Methyl 2-chloro-4-(pyrrolidin-1-yl)benzoate
OR32483-1G OR32483-5G
APOSOR32483-1GEA 7670 CZK
APOSOR32483-1G APOSOR32483-5G
Methyl-2-chloro-4-(1-pyrrolidinyl)benzenecarboxylate 95%
Methyl-2-chloro-4-(1-pyrrolidinyl)benzenecarboxylate
Vzorec: C₁₂H₁₄ClNO₂
MW: 239.7 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03844850
CAS číslo: 175153-38-5