Tisk…

1-(5-Bromo-2-thienyl)-3-(dimethylamino)-2-propen-1-one 95%

Dodavatel: Apollo Scientific

Synonyms: (E)-1-(5-Bromothiophen-2-yl)-3-(dimethylamino)prop-2-en-1-one

OR32472-1G OR32472-25G OR32472-5G
APOSOR32472-1GEA 5130 CZK
APOSOR32472-1G APOSOR32472-25G APOSOR32472-5G
1-(5-Bromo-2-thienyl)-3-(dimethylamino)-2-propen-1-one 95%
1-(5-Bromo-2-thienyl)-3-(dimethylamino)-2-propen-1-one
Vzorec: C₉H₁₀BrNOS
Storage Temperature: Ambient
MDL Number: MFCD00098108
CAS číslo: 938176-46-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR