3-Oxo-3-(4-phenylpiperazino)propanenitrile 95%
Dodavatel: Apollo Scientific
OR32458-1G
OR32458-5G
APOSOR32458-1GEA
5130
CZK
APOSOR32458-1G
APOSOR32458-5G
3-Oxo-3-(4-phenylpiperazino)propanenitrile 95%
3-Oxo-3-(4-phenylpiperazino)propanenitrile
Vzorec:
C₁₃H₁₅N₃O MW: 229,28 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01412243 CAS číslo: 14761-40-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...