2-Bromo-5-nitro-3-picoline
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Bromo-3-methyl-5-nitropyridine
OR3243-5G
OR3243-100G
OR3243-25G
APOSOR3243-5GEA
622
CZK
APOSOR3243-5G
APOSOR3243-100G
APOSOR3243-25G
2-Bromo-5-nitro-3-picoline
2-Bromo-5-nitro-3-picoline
Vzorec:
C₆H₅BrN₂O₂ MW: 217.02 Bod tání: 55…59 °C Storage Temperature: Ambient |
MDL Number:
MFCD03095065 CAS číslo: 23132-21-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...