Tisk…

4-(3,4-Dimethoxyphenyl)-3-methyl-1H-pyrazole 95%

Dodavatel: Apollo Scientific

OR32394-1G OR32394-5G
APOSOR32394-1GEA 4630 CZK
APOSOR32394-1G APOSOR32394-5G
4-(3,4-Dimethoxyphenyl)-3-methyl-1H-pyrazole 95%
4-(3,4-Dimethoxyphenyl)-3-methyl-1H-pyrazole
Vzorec: C₁₂H₁₄N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00112345
CAS číslo: 321526-09-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR