1-(2-Chloroethyl)-1H-pyrazole 97%
Dodavatel: Apollo Scientific
OR32380-25G
OR32380-5G
APOSOR32380-25GEA
10090
CZK
APOSOR32380-25G
APOSOR32380-5G
1-(2-Chloroethyl)-1H-pyrazole 97%
1-(2-Chloroethyl)-1H-pyrazole
Vzorec:
C₅H₇ClN₂ Storage Temperature: Ambient |
MDL Number:
MFCD00173067 CAS číslo: 96450-53-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...