Tisk…

2-Chloro-5-(1H-imidazol-1-ylmethyl)pyridine 95%

Dodavatel: Apollo Scientific

OR32355-1G OR32355-5G
APOSOR32355-1GEA 5130 CZK
APOSOR32355-1G APOSOR32355-5G
2-Chloro-5-(1H-imidazol-1-ylmethyl)pyridine 95%
2-Chloro-5-(1H-imidazol-1-ylmethyl)pyridine
Vzorec: C₉H₈ClN₃
MW: 193,64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04124604
CAS číslo: 230617-61-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR