2-Chloro-N-ethyl-N-phenylacetamide 95%
Dodavatel: Apollo Scientific
OR32352-1G
APOSOR32352-1GEA
5130
CZK
APOSOR32352-1G
2-Chloro-N-ethyl-N-phenylacetamide 95%
2-Chloro-N-ethyl-N-phenylacetamide
Vzorec:
C₁₀H₁₂ClNO Storage Temperature: Ambient |
MDL Number:
MFCD00227876 CAS číslo: 39086-61-8 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...