Tisk…

Ethyl pyrrolo[1,2-c]pyrimidine-3-carboxylate 95%

Dodavatel: Apollo Scientific

OR32327-1G OR32327-5G
APOSOR32327-1GEA 3440 CZK
APOSOR32327-1G APOSOR32327-5G
Ethyl pyrrolo[1,2-c]pyrimidine-3-carboxylate 95%
Ethyl pyrrolo[1,2-c]pyrimidine-3-carboxylate
Vzorec: C₁₀H₁₀N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD02082826
CAS číslo: 107407-80-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR