3,4-Dibromo-1H-pyrrole-2,5-dione 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
3,4-Dibromomaleimide
OR323258-5G
OR323258-25G
APOSOR323258-5GEA
2030
CZK
APOSOR323258-5G
APOSOR323258-25G
3,4-Dibromo-1H-pyrrole-2,5-dione 98%
3,4-Dibromo-1H-pyrrole-2,5-dione
Vzorec:
C4HBr2NO2 MW: 254,8600 g/mol Bod tání: 228…231 °C Storage Temperature: Ambient |
MDL Number:
MFCD00185696 CAS číslo: 1122-10-7 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...