3-Methyl-3,4,7,8-tetrahydro-2,5(1H,6H)-quinolinedione 95%
Dodavatel: Apollo Scientific
Synonyms:
3-Methyl-1,2,3,4,5,6,7,8-octahydroquinoline-2,5-dione
OR32280-1G
OR32280-5G
APOSOR32280-1GEA
7670
CZK
APOSOR32280-1G
APOSOR32280-5G
3-Methyl-3,4,7,8-tetrahydro-2,5(1H,6H)-quinolinedione 95%
3-Methyl-3,4,7,8-tetrahydro-2,5(1H,6H)-quinolinedione
Vzorec:
C₁₀H₁₃NO₂ MW: 179,22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13184135 CAS číslo: 77903-18-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...