7-Bromoquinazoline-2,4-diamine
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2,4-Diamino-7-bromoquinazoline, DHFR-IN-3
OR322728-1G
OR322728-250MG
OR322728-5G
APOSOR322728-1GEA
3640
CZK
APOSOR322728-1G
APOSOR322728-250MG
APOSOR322728-5G
7-Bromoquinazoline-2,4-diamine
7-Bromoquinazoline-2,4-diamine
Vzorec:
C₈H₇BrN₄ MW: 239,0720 g/mol Bod tání: 255…256 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08706214 CAS číslo: 137553-43-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...