[3-(2-Pyrimidinyloxy)phenyl]methanol 95%
Dodavatel: Apollo Scientific
OR32256-1G
OR32256-5G
APOSOR32256-1GEA
7670
CZK
APOSOR32256-1G
APOSOR32256-5G
[3-(2-Pyrimidinyloxy)phenyl]methanol 95%
[3-(2-Pyrimidinyloxy)phenyl]methanol
Vzorec:
C₁₁H₁₀N₂O₂ MW: 202,21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03791155 CAS číslo: 82440-25-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...