Tisk…

2-Aminopyrimidine-5-carbaldehyde

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Aminopyrimidine-5-carboxaldehyde

OR322504-5G
APOSOR322504-5GEA 2690 CZK
APOSOR322504-5G
2-Aminopyrimidine-5-carbaldehyde
2-Aminopyrimidine-5-carbaldehyde
Vzorec: C₅H₅N₃O
MW: 123,11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03701714
CAS číslo: 120747-84-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR