4-(Chloromethyl)-5-methyl-2-phenyloxazole
Dodavatel: Apollo Scientific
OR322355-250MG
OR322355-1G
APOSOR322355-250MGEA
3810
CZK
APOSOR322355-250MG
APOSOR322355-1G
4-(Chloromethyl)-5-methyl-2-phenyloxazole
4-(Chloromethyl)-5-methyl-2-phenyloxazole
Vzorec:
C₁₁H₁₀ClNO MW: 207,660 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00800940 CAS číslo: 103788-61-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...