Tisk…
3-Hydroxy-7-methoxy-2-naphthoic acid
Dodavatel: Apollo Scientific
Synonyms: 3-Hydroxy-7-methoxy-2-naphthalenecarboxylic Acid
OR322304-5G OR322304-1G
APOSOR322304-5GEA 1040 CZK
APOSOR322304-5G APOSOR322304-1G
3-Hydroxy-7-methoxy-2-naphthoic acid
3-Hydroxy-7-methoxy-2-naphthoic acid
Vzorec: C₁₂H₁₀O₄
MW: 218,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00059082
CAS číslo: 143355-56-0