Tisk…

4-Hydroxy-3-methylbenzaldehyde 99%

Dodavatel: Apollo Scientific
Varování

OR322238-1G OR322238-25G OR322238-5G
APOSOR322238-1GEA 705 CZK
APOSOR322238-1G APOSOR322238-25G APOSOR322238-5G
4-Hydroxy-3-methylbenzaldehyde 99%
4-Hydroxy-3-methylbenzaldehyde
Vzorec: C₈H₈O₂
Storage Temperature: Ambient
MDL Number: MFCD00012360
CAS číslo: 15174-69-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR