Tisk…

4-(Chloromethyl)benzyl alcohol

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: p-hydroxymethylbenzyl chloride, (4-(Chloromethyl)phenyl)methanol, 4-(hydroxymethyl)benzyl chloride, Benzenemethanol,4-(chloromethyl)-

OR322170-1G OR322170-25G OR322170-5G
APOSOR322170-1GEA 622 CZK
APOSOR322170-1G APOSOR322170-25G APOSOR322170-5G
4-(Chloromethyl)benzyl alcohol
4-(Chloromethyl)benzyl alcohol
Vzorec: C₈H₉ClO
MW: 156,609 g/mol
Bod tání: 58…60 °C
Storage Temperature: Ambient
MDL Number: MFCD03427011
CAS číslo: 16473-35-1
UN: 1759
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR