Tisk…

3-Amino-6-chloropyridine-2-carboxamide

Dodavatel: Apollo Scientific

OR322132-1G OR322132-25G OR322132-5G
APOSOR322132-1GEA 0 CZK
APOSOR322132-1G APOSOR322132-25G APOSOR322132-5G
3-Amino-6-chloropyridine-2-carboxamide
3-Amino-6-chloropyridine-2-carboxamide
Vzorec: C₆H₆ClN₃O
Storage Temperature: Ambient
MDL Number: MFCD09750671
CAS číslo: 175358-01-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR