N,N'-Dimethylethylendiamine 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR322130-25G
OR322130-5G
OR322130-100G
APOSOR322130-25GEA
870
CZK
APOSOR322130-25G
APOSOR322130-5G
APOSOR322130-100G
N,N'-Dimethylethylendiamine 95%
N,N'-Dimethylethylendiamine
Vzorec:
C₄H₁₂N₂ Bod varu: 118…119 °C Hustota: 0,818 g/cm³ (25 °C) Bod vzplanutí: 26 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008290 CAS číslo: 110-70-3 EINECS: 230-793-3 UN: 2734 ADR: 8+3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...