Ethyl-5-methylindole-2-carboxylate 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR322120-10G
OR322120-5G
OR322120-1G
APOSOR322120-10GEA
3910
CZK
APOSOR322120-10G
APOSOR322120-5G
APOSOR322120-1G
Ethyl-5-methylindole-2-carboxylate 97%
Ethyl-5-methylindole-2-carboxylate
Vzorec:
C₁₂H₁₃NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00022703 CAS číslo: 16382-15-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...