Tisk…

4-Nitrophenyl isothiocyanate

Dodavatel: Apollo Scientific

OR322064-5G OR322064-1G
APOSOR322064-5GEA 1620 CZK
APOSOR322064-5G APOSOR322064-1G
4-Nitrophenyl isothiocyanate
4-Nitrophenyl isothiocyanate
Vzorec: C₇H₄N₂O₂S
MW: 180,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007307
CAS číslo: 2131-61-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR