4,6-Dimethyl-2-hydroxypyrimidine 97%
Dodavatel: Apollo Scientific
Synonyms:
2-Hydroxy-4,6-dimethylpyrimidine, 4,6-Dimethyl-2-pyrimidinol, 4,6-Dimethylpyrimidin-2(1H)-one
OR322007-100G
APOSOR322007-100GEA
1870
CZK
APOSOR322007-100G
4,6-Dimethyl-2-hydroxypyrimidine 97%
4,6-Dimethyl-2-hydroxypyrimidine
Vzorec:
C₆H₈N₂O MW: 124,14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00006072 CAS číslo: 108-79-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...