3-(Benzyloxy)-2-methyl-4H-pyran-4-one 95%
Dodavatel: Apollo Scientific
OR32197-1G
OR32197-5G
APOSOR32197-1GEA
6390
CZK
APOSOR32197-1G
APOSOR32197-5G
3-(Benzyloxy)-2-methyl-4H-pyran-4-one 95%
3-(Benzyloxy)-2-methyl-4H-pyran-4-one
Vzorec:
C₁₃H₁₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD00207271 CAS číslo: 61049-69-2 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...