Tisk…

Methyl-5-methyl-3-phenyl-4-isoxazolecarboxylate 95%

Dodavatel: Apollo Scientific

OR32169-1G OR32169-5G
APOSOR32169-1GEA 9560 CZK
APOSOR32169-1G APOSOR32169-5G
Methyl-5-methyl-3-phenyl-4-isoxazolecarboxylate 95%
Methyl-5-methyl-3-phenyl-4-isoxazolecarboxylate
Vzorec: C₁₂H₁₁NO₃
MW: 217,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00119105
CAS číslo: 2065-28-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR