Tisk…

Methyl-4-(1H-imidazol-1-yl)benzenecarboxylate 98%

Dodavatel: Apollo Scientific

Synonyms: Methyl 4-(1H-imidazol-1-yl)benzoate

OR32165-1G
APOSOR32165-1GEA 870 CZK
APOSOR32165-1G
Methyl-4-(1H-imidazol-1-yl)benzenecarboxylate 98%
Methyl-4-(1H-imidazol-1-yl)benzenecarboxylate
Vzorec: C₁₁H₁₀N₂O₂
MW: 202,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00127080
CAS číslo: 101184-08-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR