2-(1H-Imidazol-1-yl)benzenecarbonitrile 95%
Dodavatel: Apollo Scientific
OR32163-25G
OR32163-5G
APOSOR32163-25GEA
13390
CZK
APOSOR32163-25G
APOSOR32163-5G
2-(1H-Imidazol-1-yl)benzenecarbonitrile 95%
2-(1H-Imidazol-1-yl)benzenecarbonitrile
Vzorec:
C₁₀H₇N₃ MW: 169,19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00127074 CAS číslo: 25373-49-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...