Tisk…

3-Chloro-2,2-dimethyl-N-phenylpropanamide 95%

Dodavatel: Apollo Scientific

OR32156-1G OR32156-5G
APOSOR32156-1GEA 7670 CZK
APOSOR32156-1G APOSOR32156-5G
3-Chloro-2,2-dimethyl-N-phenylpropanamide 95%
3-Chloro-2,2-dimethyl-N-phenylpropanamide
Vzorec: C₁₁H₁₄ClNO
Storage Temperature: Ambient
MDL Number: MFCD07698577
CAS číslo: 82820-74-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR