Boc-(2S,5R)-5-phenylpyrrolidine-2-carboxylic acid
Dodavatel: Apollo Scientific
Synonyms:
(2S,5R)-1-(tert-Butoxycarbonyl)-5-phenylpyrrolidine-2-carboxylic acid
OR321545-250MG
APOSOR321545-250MGEA
8500
CZK
APOSOR321545-250MG
Boc-(2S,5R)-5-phenylpyrrolidine-2-carboxylic acid
Boc-(2S,5R)-5-phenylpyrrolidine-2-carboxylic acid
Vzorec:
C₁₆H₂₁NO₄ MW: 291,35 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD01321008 CAS číslo: 221352-49-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...