(R)-1-Boc-3-Benzylpiperazine
Dodavatel: Apollo Scientific
OR321498-1G
OR321498-250MG
APOSOR321498-1GEA
5590
CZK
APOSOR321498-1G
APOSOR321498-250MG
(R)-1-Boc-3-Benzylpiperazine
(R)-1-Boc-3-Benzylpiperazine
Vzorec:
C₁₆H₂₄N₂O₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD07772103 CAS číslo: 947272-49-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...