(3R,4R)-1-Benzyl-N-4-dimethylpiperidin-3-amine 99%
Dodavatel: Apollo Scientific
Synonyms:
(3R,4R)-1-Benzyl-N,4-dimethylpiperidin-3-amine
OR321454-100G
OR321454-5G
OR321454-25G
APOSOR321454-100GEA
19910
CZK
APOSOR321454-100G
APOSOR321454-5G
APOSOR321454-25G
(3R,4R)-1-Benzyl-N-4-dimethylpiperidin-3-amine 99%
(3R,4R)-1-Benzyl-N-4-dimethylpiperidin-3-amine
Vzorec:
C₁₄H₂₂N₂ MW: 218.3379 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09475548 CAS číslo: 477600-70-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...