Tisk…

3-Chloro-2-pyrazinemethanol 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: (3-Chloropyrazin-2-yl)methanol

OR321372-1G OR321372-5G OR321372-10G
APOSOR321372-1GEA 2480 CZK
APOSOR321372-1G APOSOR321372-5G APOSOR321372-10G
3-Chloro-2-pyrazinemethanol 97%
3-Chloro-2-pyrazinemethanol
Vzorec: C₅H₅ClN₂O
MW: 144,56 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11977628
CAS číslo: 89283-32-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR