Tisk…

tert-Butyl piperazin-1-ylcarbamate 97%

Dodavatel: Apollo Scientific
Varování

OR321350-250MG
APOSOR321350-250MGEA 2700 CZK
APOSOR321350-250MG
tert-Butyl piperazin-1-ylcarbamate 97%
tert-Butyl piperazin-1-ylcarbamate
Vzorec: C₉H₁₉N₃O₂
MW: 201,2661 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11840387
CAS číslo: 147081-80-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR