3-(4-Methoxyphenyl)piperidine 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR321331-1G
OR321331-100MG
OR321331-250MG
APOSOR321331-1GEA
11210
CZK
APOSOR321331-1G
APOSOR321331-100MG
APOSOR321331-250MG
3-(4-Methoxyphenyl)piperidine 97%
3-(4-Methoxyphenyl)piperidine
Vzorec:
C₁₂H₁₇NO Storage Temperature: Refrigerator |
MDL Number:
MFCD09837353 CAS číslo: 19725-26-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...