Tisk…

3,6-Dichloropyrazine-2-carbonitrile 97%

Dodavatel: Apollo Scientific
Nebezpečí

OR321323-1G OR321323-250MG
APOSOR321323-1GEA 3940 CZK
APOSOR321323-1G APOSOR321323-250MG
3,6-Dichloropyrazine-2-carbonitrile 97%
3,6-Dichloropyrazine-2-carbonitrile
Vzorec: C₅HCl₂N₃
Storage Temperature: Ambient
MDL Number: MFCD22199269
CAS číslo: 356783-16-9
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR