Tisk…

(R)-1-Boc-3-Propylpiperazine 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: tert-Butyl (R)-3-propylpiperazine-1-carboxylate

OR321301-1G OR321301-5G
APOSOR321301-1GEA 2030 CZK
APOSOR321301-1G APOSOR321301-5G
(R)-1-Boc-3-Propylpiperazine 95%
(R)-1-Boc-3-Propylpiperazine
Vzorec: C₁₂H₂₄N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD07772107
CAS číslo: 928025-57-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR