7-Hydroxy-6-propyl-2,3-dihydro-1H,5H-pyrido[3,2,1-ij]quinolin-5-one 95%
Dodavatel: Apollo Scientific
Synonyms:
4-Hydroxy-3-propyl-1-azatricyclo[7.3.1.05,13]trideca-3,5(13),6,8-tetraen-2-one
OR32126-1G
OR32126-5G
APOSOR32126-1GEA
5130
CZK
APOSOR32126-1G
APOSOR32126-5G
7-Hydroxy-6-propyl-2,3-dihydro-1H,5H-pyrido[3,2,1-ij]quinolin-5-one 95%
7-Hydroxy-6-propyl-2,3-dihydro-1H,5H-pyrido[3,2,1-ij]quinolin-5-one
Vzorec:
C₁₅H₁₇NO₂ MW: 243,31 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03791142 CAS číslo: 57625-51-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...