Tisk…

7-Methoxybenzofuran-2-carboxylic acid 95%

Dodavatel: Apollo Scientific

OR32111-5G
APOSOR32111-5GEA 1780 CZK
APOSOR32111-5G
7-Methoxybenzofuran-2-carboxylic acid 95%
7-Methoxybenzofuran-2-carboxylic acid
Vzorec: C₁₀H₈O₄
MW: 192,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00060516
CAS číslo: 4790-79-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR