1-[(2-Chloro-1,3-thiazol-5-yl)methyl]-4-methylpiperazine 95%
Dodavatel: Apollo Scientific
OR32089-1G
OR32089-5G
APOSOR32089-1GEA
7670
CZK
APOSOR32089-1G
APOSOR32089-5G
1-[(2-Chloro-1,3-thiazol-5-yl)methyl]-4-methylpiperazine 95%
1-[(2-Chloro-1,3-thiazol-5-yl)methyl]-4-methylpiperazine
Vzorec:
C₉H₁₄ClN₃S Storage Temperature: Ambient |
MDL Number:
MFCD01568175 CAS číslo: 341967-70-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...