Tisk…

3-Chloro-N-[4-(dimethylamino)phenyl]-2,2-dimethylpropanamide 95%

Dodavatel: Apollo Scientific

OR32082-1G OR32082-25G OR32082-5G
APOSOR32082-1GEA 5130 CZK
APOSOR32082-1G APOSOR32082-25G APOSOR32082-5G
3-Chloro-N-[4-(dimethylamino)phenyl]-2,2-dimethylpropanamide 95%
3-Chloro-N-[4-(dimethylamino)phenyl]-2,2-dimethylpropanamide
Vzorec: C₁₃H₁₉ClN₂O
Storage Temperature: Ambient
MDL Number: MFCD00793368
CAS číslo: 341966-00-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR