2-(Bromomethyl)-2,6-dihydroimidazo[1,2-c]quinazolin-5(3H)-one 95%
Dodavatel: Apollo Scientific
OR32051-1G
OR32051-5G
APOSOR32051-1GEA
15310
CZK
APOSOR32051-1G
APOSOR32051-5G
2-(Bromomethyl)-2,6-dihydroimidazo[1,2-c]quinazolin-5(3H)-one 95%
2-(Bromomethyl)-2,6-dihydroimidazo[1,2-c]quinazolin-5(3H)-one
Vzorec:
C₁₁H₁₀BrN₃O Storage Temperature: Ambient |
MDL Number:
MFCD08056599 CAS číslo: 139047-54-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...