Tisk…

1-(4-Acetylphenyl)-4-piperidinecarboxylic acid 95%

Dodavatel: Apollo Scientific

OR32034-1G OR32034-5G
APOSOR32034-1GEA 7670 CZK
APOSOR32034-1G APOSOR32034-5G
1-(4-Acetylphenyl)-4-piperidinecarboxylic acid 95%
1-(4-Acetylphenyl)-4-piperidinecarboxylic acid
Vzorec: C₁₄H₁₇NO₃
MW: 247.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00231449
CAS číslo: 250713-76-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR